Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054094.5 | NP_473435.1 | 7 - 8 | 1064 | 75 | |
GenBank mRNA | AB059428.1 | - | 7 - 8 | 1068 | 75 | |
AK149550.1 | - | 7 - 8 | 1074 | 75 | ||
AK149586.1 | - | 7 - 8 | 1090 | 75 | ||
BC016414.1 | - | 7 - 8 | 1061 | 4238964 | 75 | |
BC022149.1 | - | 7 - 8 | 1042 | 5102652 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |