Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290371.1 | NP_001277300.1 | 5 - 6 | 578 | 71 | |
NM_009666.4 | NP_033796.1 | 5 - 6 | 650 | 71 | ||
GenBank mRNA | AI323496.1 | - | 1 - 2 | 71 | ||
AK029358.1 | - | 5 - 6 | 654 | 71 | ||
D31768.1 | - | 5 - 6 | 641 | 71 | ||
D31769.1 | - | 5 - 6 | 569 | 71 | ||
KC731591.1 | - | 5 - 6 | 578 | 71 | ||
M10095.1 | - | 2 - 3 | 458 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons |