Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011077.2 | NP_035207.1 | 19 - 20 | 2495 | 98 | |
GenBank mRNA | AK158481.1 | - | 19 - 20 | 2522 | 98 | |
BC137925.1 | - | 19 - 20 | 2242 | 8860944 | 98 | |
BC137926.1 | - | 19 - 20 | 2242 | 8860945 | 98 | |
EF194891.1 | - | 19 - 20 | 2029 | 98 | ||
U49908.1 | - | 19 - 20 | 2487 | 98 | ||
U73910.1 | - | 19 - 20 | 2495 | 98 | ||
U75646.1 | - | 19 - 20 | 2076 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |