Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011077.2 | NP_035207.1 | 20 - 21 | 2595 | 68 | |
GenBank mRNA | AK158481.1 | - | 20 - 21 | 2622 | 68 | |
BC137925.1 | - | 20 - 21 | 2342 | 8860944 | 68 | |
BC137926.1 | - | 20 - 21 | 2342 | 8860945 | 68 | |
EF194891.1 | - | 20 - 21 | 2129 | 68 | ||
U49908.1 | - | 20 - 21 | 2587 | 68 | ||
U73910.1 | - | 20 - 21 | 2595 | 68 | ||
U75646.1 | - | 20 - 21 | 2176 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |