Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phex | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011077.2 | NP_035207.1 | 3 - 4 | 879 | 66 | |
GenBank mRNA | AK158481.1 | - | 3 - 4 | 906 | 66 | |
BC137925.1 | - | 3 - 4 | 626 | 8860944 | 66 | |
BC137926.1 | - | 3 - 4 | 626 | 8860945 | 66 | |
EF194891.1 | - | 3 - 4 | 413 | 66 | ||
U49908.1 | - | 3 - 4 | 871 | 66 | ||
U73910.1 | - | 3 - 4 | 879 | 66 | ||
U75646.1 | - | 3 - 4 | 460 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |