Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081253.1 | NP_001074722.1 | 3 - 4 | 2003 | 84 | |
GenBank mRNA | AK019714.1 | - | 3 - 4 | 1941 | 84 | |
BC026503.1 | - | 2 - 3 | 297 | 84 | ||
BC098484.1 | - | 3 - 4 | 2003 | 30053208 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |