Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175105.3 | NP_780314.1 | 1 - 2 | 69 | ||
GenBank mRNA | AB028148.1 | - | 1 - 2 | 69 | ||
AK006005.1 | - | 1 - 2 | 69 | |||
AK075757.1 | - | 1 - 2 | 69 | |||
AK137326.1 | - | 1 - 2 | 69 | |||
BC037088.1 | - | 1 - 2 | 662 | 69 | ||
BC068103.1 | - | 2 - 3 | 69 | |||
BC115612.2 | - | 2 - 3 | 69 | |||
BC115613.2 | - | 2 - 3 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |