Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289787.1 | NP_001276716.1 | 4 - 5 | 619 | 73 | |
NM_001289788.1 | NP_001276717.1 | 4 - 5 | 619 | 73 | ||
NM_030727.5 | NP_109652.3 | 6 - 7 | 805 | 73 | ||
GenBank mRNA | AF529192.1 | - | 6 - 7 | 805 | 73 | |
AY024359.2 | - | 4 - 5 | 571 | 73 | ||
BC108986.1 | - | 4 - 5 | 611 | 40047724 | 73 | |
BC108987.1 | - | 4 - 5 | 611 | 40047725 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |