Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172827.3 | NP_766415.1 | 2 - 3 | 943 | 79 | |
GenBank mRNA | AK029094.1 | - | 2 - 3 | 944 | 79 | |
AK036967.1 | - | 2 - 3 | 944 | 79 | ||
AK161484.1 | - | 170 | 79 | |||
BC066791.1 | - | 3 - 4 | 1267 | 30054751 | 79 | |
BC120925.1 | - | 2 - 3 | 900 | 40098425 | 79 | |
BC120926.1 | - | 2 - 3 | 900 | 40098428 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |