Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172827.3 | NP_766415.1 | 7 - 8 | 1593 | 100 | |
GenBank mRNA | AK029094.1 | - | 7 - 8 | 1594 | 100 | |
AK036967.1 | - | 7 - 8 | 1594 | 100 | ||
AK161484.1 | - | 820 | 100 | |||
BC066791.1 | - | 8 - 9 | 1917 | 30054751 | 100 | |
BC120925.1 | - | 7 - 8 | 1550 | 40098425 | 100 | |
BC120926.1 | - | 7 - 8 | 1550 | 40098428 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |