Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700035081.mm.1 |
NM_001033173.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035081.mm.1 |
NM_001033173.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035081.mm.1 |
TC0700004215.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700035082.mm.1 |
NM_001033173.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035082.mm.1 |
NM_001033173.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035082.mm.1 |
TC0700004215.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700035083.mm.1 |
NM_001033173.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035083.mm.1 |
NM_001033173.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700035083.mm.1 |
TC0700004215.mm.1 |
MTA TC |