Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033173.1 | NP_001028345.1 | 7 - 8 | 1393 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Protease Mus musculus Probe spans exons |