Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahcyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171000.2 | NP_001164471.2 | 14 - 15 | 1731 | 70 | |
NM_001171001.1 | NP_001164472.1 | 14 - 15 | 1451 | 70 | ||
NM_021414.6 | NP_067389.5 | 14 - 15 | 1734 | 70 | ||
GenBank mRNA | AK014539.1 | - | 15 - 16 | 1685 | 70 | |
AK053527.1 | - | 14 - 15 | 1415 | 70 | ||
AK122382.1 | - | 12 - 13 | 1437 | 70 | ||
AK161892.1 | - | 3 - 4 | 211 | 70 | ||
BC037447.1 | - | 2 - 3 | 178 | 70 | ||
BC051504.1 | - | 4 - 5 | 364 | 70 | ||
BC059255.1 | - | 10 - 11 | 906 | 70 | ||
BC079660.1 | - | 14 - 15 | 1644 | 6853550 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |