Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xylt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175645.3 | NP_783576.2 | 4 - 5 | 1275 | 87 | |
GenBank mRNA | AK033309.1 | - | 4 - 5 | 1277 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |