Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xylt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175645.3 | NP_783576.2 | 9 - 10 | 2208 | 89 | |
GenBank mRNA | BM964190.1 | - | 1 - 2 | 172 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |