Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013903.2 | NP_038931.1 | 8 - 9 | 1260 | 120 | |
GenBank mRNA | AF155933.1 | - | 8 - 9 | 1260 | 120 | |
BC152335.1 | - | 8 - 9 | 1264 | 40142709 | 120 | |
BC152336.1 | - | 8 - 9 | 1264 | 40142712 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |