Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008611.4 | NP_032637.3 | 8 - 9 | 1297 | 95 | |
GenBank mRNA | AK089234.1 | - | 8 - 9 | 1268 | 95 | |
AK089336.1 | - | 8 - 9 | 1272 | 95 | ||
AK089516.1 | - | 8 - 9 | 1258 | 95 | ||
AK137468.1 | - | 8 - 9 | 1269 | 95 | ||
AK154937.1 | - | 8 - 9 | 1268 | 95 | ||
AK155832.1 | - | 8 - 9 | 1268 | 95 | ||
AK155901.1 | - | 8 - 9 | 1272 | 95 | ||
AK170388.1 | - | 8 - 9 | 1270 | 95 | ||
AK170760.1 | - | 8 - 9 | 1268 | 95 | ||
AK171168.1 | - | 8 - 9 | 1268 | 95 | ||
AK171613.1 | - | 8 - 9 | 1270 | 95 | ||
BC042742.1 | - | 8 - 9 | 1278 | 5032770 | 95 | |
Y13342.1 | - | 8 - 9 | 1195 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |