Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019471.3 | NP_062344.1 | 4 - 5 | 684 | 66 | |
GenBank mRNA | AK020292.1 | - | 4 - 5 | 685 | 66 | |
AY622970.1 | - | 1 - 2 | 103 | 66 | ||
BC130027.1 | - | 4 - 5 | 676 | 40140858 | 66 | |
Y13185.1 | - | 4 - 5 | 673 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |