Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281934.1 | NP_001268863.1 | 3 - 4 | 1331 | 78 | |
NM_008794.2 | NP_032820.2 | 3 - 4 | 1067 | 78 | ||
NR_104055.1 | 3 - 4 | 1067 | 78 | |||
GenBank mRNA | AK148416.1 | - | 3 - 4 | 652 | 78 | |
AK151980.1 | - | 3 - 4 | 654 | 78 | ||
AK165389.1 | - | 3 - 4 | 673 | 78 | ||
BC006730.1 | - | 2 - 3 | 568 | 3962441 | 78 | |
BC007322.1 | - | 3 - 4 | 930 | 78 | ||
BC056456.1 | - | 3 - 4 | 1067 | 78 | ||
U48830.1 | - | 3 - 4 | 639 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |