Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc37a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293630.1 | NP_001280559.1 | 1 - 2 | 357 | 87 | |
GenBank mRNA | AK155339.1 | - | 1 - 2 | 354 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Transporter Mus musculus Probe spans exons |