Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008605.3 | NP_032631.3 | 5 - 6 | 853 | 83 | |
GenBank mRNA | AK089309.1 | - | 5 - 6 | 822 | 83 | |
AK089452.1 | - | 5 - 6 | 822 | 83 | ||
AK089509.1 | - | 6 - 7 | 696 | 83 | ||
AK089523.1 | - | 5 - 6 | 822 | 83 | ||
AK090051.1 | - | 5 - 6 | 824 | 83 | ||
AK150596.1 | - | 5 - 6 | 822 | 83 | ||
AK151021.1 | - | 5 - 6 | 822 | 83 | ||
AK151273.1 | - | 5 - 6 | 822 | 83 | ||
AK151933.1 | - | 5 - 6 | 822 | 83 | ||
AK155139.1 | - | 5 - 6 | 821 | 83 | ||
AK155743.1 | - | 5 - 6 | 822 | 83 | ||
AK155747.1 | - | 5 - 6 | 822 | 83 | ||
AK155748.1 | - | 5 - 6 | 822 | 83 | ||
AK155841.1 | - | 5 - 6 | 880 | 83 | ||
AK155865.1 | - | 5 - 6 | 821 | 83 | ||
AK170360.1 | - | 5 - 6 | 822 | 83 | ||
AK170364.1 | - | 5 - 6 | 822 | 83 | ||
AK170500.1 | - | 6 - 7 | 696 | 83 | ||
AK170667.1 | - | 4 - 5 | 1869 | 83 | ||
AK170798.1 | - | 5 - 6 | 821 | 83 | ||
AK170804.1 | - | 5 - 6 | 821 | 83 | ||
AK170976.1 | - | 5 - 6 | 822 | 83 | ||
AK171006.1 | - | 5 - 6 | 822 | 83 | ||
AK171032.1 | - | 5 - 6 | 821 | 83 | ||
AK171036.1 | - | 5 - 6 | 822 | 83 | ||
AK171176.1 | - | 5 - 6 | 822 | 83 | ||
AK171428.1 | - | 5 - 6 | 822 | 83 | ||
AK171435.1 | - | 4 - 5 | 1869 | 83 | ||
AK171447.1 | - | 5 - 6 | 822 | 83 | ||
AK171448.1 | - | 5 - 6 | 822 | 83 | ||
AK171449.1 | - | 5 - 6 | 822 | 83 | ||
AK171450.1 | - | 5 - 6 | 822 | 83 | ||
AK171466.1 | - | 5 - 6 | 822 | 83 | ||
AK171468.1 | - | 5 - 6 | 822 | 83 | ||
AK171470.1 | - | 5 - 6 | 822 | 83 | ||
AK171472.1 | - | 5 - 6 | 822 | 83 | ||
AK171485.1 | - | 5 - 6 | 822 | 83 | ||
AK171518.1 | - | 5 - 6 | 822 | 83 | ||
AK171542.1 | - | 5 - 6 | 822 | 83 | ||
AK171570.1 | - | 5 - 6 | 822 | 83 | ||
BC019135.1 | - | 5 - 6 | 5037674 | 83 | ||
CT010377.1 | - | 5 - 6 | 83 | |||
M82831.1 | - | 5 - 6 | 769 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |