Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016803.3 | NP_058083.2 | 2 - 3 | 511 | 81 | |
GenBank mRNA | CJ142827.1 | - | 2 - 3 | 514 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |