Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027425.3 | NP_081701.2 | 3 - 4 | 398 | 104 | |
GenBank mRNA | AK011847.1 | - | 3 - 4 | 368 | 104 | |
AK083205.1 | - | 3 - 4 | 383 | 104 | ||
AK173195.1 | - | 3 - 4 | 359 | 104 | ||
BC017645.1 | - | 3 - 4 | 360 | 104 | ||
BC038265.1 | - | 3 - 4 | 339 | 104 | ||
BC137736.1 | - | 3 - 4 | 347 | 8860755 | 104 | |
BC137737.1 | - | 3 - 4 | 347 | 8860756 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |