Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081127.1 | NP_001074596.1 | 13 - 14 | 2220 | 71 | |
GenBank mRNA | BC157953.1 | - | 13 - 14 | 2055 | 9007283 | 71 |
BC158014.1 | - | 13 - 14 | 2053 | 9088151 | 71 | |
BC172106.1 | - | 13 - 14 | 2050 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |