Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700024058.mm.1 |
TC0700002720.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700024058.mm.1 |
NM_001082548.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024058.mm.1 |
NM_001082548.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024058.mm.1 |
NM_011464.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024058.mm.1 |
NM_011464.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024057.mm.1 |
TC0700002720.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700024057.mm.1 |
NM_001082548.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024057.mm.1 |
NM_001082548.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024057.mm.1 |
NM_011464.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024057.mm.1 |
NM_011464.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024060.mm.1 |
TC0700002720.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700024060.mm.1 |
NM_001082548.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024060.mm.1 |
NM_001082548.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024060.mm.1 |
NM_011464.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700024060.mm.1 |
NM_011464.2 |
Refseq |