Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016881.2 | NP_058577.1 | 5 - 6 | 507 | 79 | |
GenBank mRNA | AF043514.1 | - | 5 - 6 | 507 | 79 | |
AK002858.1 | - | 5 - 6 | 449 | 79 | ||
AK003334.1 | - | 5 - 6 | 465 | 79 | ||
AK005074.1 | - | 5 - 6 | 489 | 79 | ||
AK012683.1 | - | 5 - 6 | 491 | 79 | ||
AK134850.1 | - | 459 | 79 | |||
AK145402.1 | - | 5 - 6 | 459 | 79 | ||
AK149116.1 | - | 5 - 6 | 469 | 79 | ||
BC037101.1 | - | 5 - 6 | 484 | 79 | ||
BC046325.1 | - | 5 - 6 | 455 | 6395635 | 79 | |
BC094448.1 | - | 5 - 6 | 473 | 4238664 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |