Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600009516.mm.1 |
NM_001013778.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009516.mm.1 |
NM_001013778.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009516.mm.1 |
TC1600001194.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600009514.mm.1 |
NM_001013778.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009514.mm.1 |
NM_001013778.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009514.mm.1 |
TC1600001194.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600009515.mm.1 |
NM_001013778.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009515.mm.1 |
NM_001013778.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009515.mm.1 |
TC1600001194.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600009506.mm.1 |
NM_001013778.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009506.mm.1 |
NM_001013778.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009506.mm.1 |
TC1600001194.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600009513.mm.1 |
NM_001013778.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009513.mm.1 |
NM_001013778.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600009513.mm.1 |
TC1600001194.mm.1 |
MTA TC |