Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172290.3 | NP_758494.2 | 2 - 3 | 372 | 83 | |
GenBank mRNA | AF282980.1 | - | 1 - 2 | 169 | 83 | |
AK042854.1 | - | 2 - 3 | 374 | 83 | ||
AK045973.1 | - | 2 - 3 | 372 | 83 | ||
AK046377.1 | - | 2 - 3 | 372 | 83 | ||
AK138263.1 | - | 2 - 3 | 360 | 83 | ||
AK140516.1 | - | 2 - 3 | 374 | 83 | ||
AK158752.1 | - | 1 - 2 | 391 | 83 | ||
BC023307.1 | - | 1 - 2 | 212 | 4480983 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |