Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hyls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029762.1 | NP_084038.1 | 1 - 2 | 190 | 73 | |
GenBank mRNA | AK013896.1 | - | 1 - 2 | 224 | 73 | |
AK083607.1 | - | 1 - 2 | 219 | 73 | ||
AK131639.1 | - | 1 - 2 | 154 | 73 | ||
BC035953.1 | - | 1 - 2 | 166 | 4911163 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |