Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102613.1 | NP_001096083.1 | 8 - 9 | 1168 | 106 | |
GenBank mRNA | BC052555.1 | - | 3 - 4 | 472 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |