Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161623.1 | NP_001155095.1 | 11 - 12 | 995 | 110 | |
NM_009862.2 | NP_033992.2 | 13 - 14 | 1372 | 110 | ||
GenBank mRNA | AF081536.1 | - | 13 - 14 | 1372 | 110 | |
AF081537.1 | - | 12 - 13 | 1071 | 110 | ||
AF081538.1 | - | 12 - 13 | 1071 | 110 | ||
AF098068.1 | - | 12 - 13 | 1118 | 110 | ||
AK147013.1 | - | 12 - 13 | 1089 | 110 | ||
AK167850.1 | - | 11 - 12 | 983 | 110 | ||
BC006856.1 | - | 12 - 13 | 1087 | 110 | ||
BC028635.1 | - | 12 - 13 | 1125 | 1262927 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |