Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190974.1 | NP_001177903.1 | 16 - 17 | 2243 | 100 | |
NM_001190975.1 | NP_001177904.1 | 15 - 16 | 1979 | 100 | ||
NM_009465.4 | NP_033491.2 | 17 - 18 | 2270 | 100 | ||
GenBank mRNA | AK155567.1 | - | 18 - 19 | 2156 | 100 | |
AK161294.1 | - | 10 - 11 | 943 | 100 | ||
AK170010.1 | - | 16 - 17 | 2217 | 100 | ||
AK170897.1 | - | 16 - 17 | 1878 | 100 | ||
BC008551.1 | - | 12 - 13 | 1360 | 100 | ||
BC046618.1 | - | 17 - 18 | 2107 | 6494383 | 100 | |
BC050914.1 | - | 17 - 18 | 2078 | 6418252 | 100 | |
BC058230.1 | - | 17 - 18 | 2131 | 6313662 | 100 | |
X63535.1 | - | 17 - 18 | 2019 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |