Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900004122.mm.1 |
NM_175482.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004122.mm.1 |
NM_175482.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004122.mm.1 |
TC0900000589.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004118.mm.1 |
NM_175482.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004118.mm.1 |
NM_175482.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004118.mm.1 |
TC0900000589.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004120.mm.1 |
NM_175482.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004120.mm.1 |
NM_175482.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004120.mm.1 |
TC0900000589.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004119.mm.1 |
NM_175482.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004119.mm.1 |
NM_175482.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004119.mm.1 |
TC0900000589.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004117.mm.1 |
NM_175482.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004117.mm.1 |
NM_175482.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004117.mm.1 |
TC0900000589.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900004121.mm.1 |
NM_175482.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004121.mm.1 |
NM_175482.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900004121.mm.1 |
TC0900000589.mm.1 |
MTA TC |