Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpv17l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033564.3 | NP_291042.2 | 2 - 3 | 433 | 114 | |
GenBank mRNA | AF305634.1 | - | 2 - 3 | 403 | 114 | |
BC094450.1 | - | 2 - 3 | 400 | 4973897 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |