Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080457.3 | NP_536705.3 | 12 - 13 | 8926 | 62 | |
GenBank mRNA | AF218265.1 | - | 12 - 13 | 2231 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |