Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159421.1 | NP_001152893.1 | 2 - 3 | 631 | 124 | |
NM_001159422.1 | NP_001152894.1 | 3 - 4 | 408 | 124 | ||
NM_198645.2 | NP_941047.1 | 3 - 4 | 757 | 124 | ||
GenBank mRNA | AK135450.1 | - | 3 - 4 | 337 | 124 | |
AV583692.1 | - | 3 - 4 | 408 | 124 | ||
BC024406.1 | - | 3 - 4 | 321 | 124 | ||
BC024862.1 | - | 3 - 4 | 732 | 5356370 | 124 | |
BC053690.1 | - | 3 - 4 | 309 | 124 | ||
CF617453.1 | - | 3 - 4 | 735 | 124 | ||
CJ094333.1 | - | 2 - 3 | 212 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |