Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038663.1 | NP_001033752.1 | 7 - 8 | 1209 | 82 | |
NM_011949.3 | NP_036079.1 | 7 - 8 | 1209 | 82 | ||
GenBank mRNA | AK035386.1 | - | 7 - 8 | 1183 | 82 | |
AK048127.1 | - | 7 - 8 | 1211 | 82 | ||
AK087925.1 | - | 7 - 8 | 1184 | 82 | ||
AK132241.1 | - | 7 - 8 | 1197 | 82 | ||
AK148853.1 | - | 7 - 8 | 1171 | 82 | ||
AK211534.1 | - | 2 - 3 | 168 | 82 | ||
BC006708.1 | - | 2 - 3 | 255 | 82 | ||
BC058258.1 | - | 7 - 8 | 1152 | 6468233 | 82 | |
D10939.1 | - | 7 - 8 | 1024 | 82 | ||
X58712.1 | - | 7 - 8 | 982 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |