Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080999.2 | NP_001074468.1 | 2 - 3 | 920 | 58 | |
NM_001081000.2 | NP_001074469.1 | 2 - 3 | 967 | 58 | ||
NM_001195205.1 | NP_001182134.1 | 2 - 3 | 920 | 58 | ||
GenBank mRNA | AK034102.1 | - | 2 - 3 | 800 | 58 | |
AK080170.1 | - | 2 - 3 | 795 | 58 | ||
AK133844.1 | - | 2 - 3 | 795 | 58 | ||
AK134368.1 | - | 2 - 3 | 921 | 58 | ||
AK146391.1 | - | 2 - 3 | 860 | 58 | ||
AK150482.1 | - | 2 - 3 | 765 | 58 | ||
AK151311.1 | - | 2 - 3 | 765 | 58 | ||
AK151422.1 | - | 2 - 3 | 791 | 58 | ||
AK151725.1 | - | 2 - 3 | 765 | 58 | ||
AK151778.1 | - | 2 - 3 | 765 | 58 | ||
AK152055.1 | - | 2 - 3 | 765 | 58 | ||
AK152201.1 | - | 2 - 3 | 765 | 58 | ||
AK152363.1 | - | 2 - 3 | 765 | 58 | ||
AK152548.1 | - | 2 - 3 | 765 | 58 | ||
AK152903.1 | - | 2 - 3 | 765 | 58 | ||
AK153043.1 | - | 2 - 3 | 765 | 58 | ||
AK153190.1 | - | 2 - 3 | 791 | 58 | ||
AK169240.1 | - | 2 - 3 | 785 | 58 | ||
BC027431.1 | - | 1 - 2 | 1032 | 3156359 | 58 | |
BC038486.1 | - | 2 - 3 | 793 | 3155739 | 58 | |
BC046976.1 | - | 2 - 3 | 801 | 6337243 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |