Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153790.2 | NP_722485.1 | 5 - 6 | 1148 | 66 | |
GenBank mRNA | AB024432.1 | - | 5 - 6 | 1115 | 66 | |
AF522197.1 | - | 5 - 6 | 1149 | 66 | ||
AK142378.1 | - | 5 - 6 | 1116 | 66 | ||
AK143208.1 | - | 5 - 6 | 991 | 66 | ||
AK170331.1 | - | 1 - 2 | 202 | 66 | ||
BC059226.1 | - | 5 - 6 | 1080 | 66 | ||
BC060084.1 | - | 5 - 6 | 1080 | 66 | ||
BC139758.1 | - | 5 - 6 | 1101 | 40047931 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |