Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164609.1 | NP_001158081.1 | 16 - 17 | 1726 | 80 | |
NM_001164610.1 | NP_001158082.1 | 15 - 16 | 1687 | 80 | ||
NM_001164611.1 | NP_001158083.1 | 15 - 16 | 1684 | 80 | ||
NM_029945.3 | NP_084221.2 | 16 - 17 | 1774 | 80 | ||
GenBank mRNA | AK019466.2 | - | 16 - 17 | 1775 | 80 | |
AK031029.1 | - | 15 - 16 | 1678 | 80 | ||
AK031685.1 | - | 15 - 16 | 1686 | 80 | ||
AK035892.1 | - | 15 - 16 | 1679 | 80 | ||
AK036055.1 | - | 15 - 16 | 1679 | 80 | ||
AK036380.1 | - | 15 - 16 | 1683 | 80 | ||
AK129354.1 | - | 16 - 17 | 1751 | 80 | ||
AK138720.1 | - | 15 - 16 | 1661 | 80 | ||
AK164234.1 | - | 16 - 17 | 1718 | 80 | ||
AK164456.1 | - | 13 - 14 | 2152 | 80 | ||
AK183804.1 | - | 1 - 2 | 157 | 80 | ||
BC026767.1 | - | 15 - 16 | 1660 | 5137008 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |