Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134399.1 | NP_001127871.1 | 12 - 13 | 1783 | 104 | |
NM_172522.4 | NP_766110.3 | 11 - 12 | 1690 | 104 | ||
GenBank mRNA | AK032661.1 | - | 11 - 12 | 1502 | 104 | |
AK051642.1 | - | 11 - 12 | 1627 | 104 | ||
AK122555.2 | - | 12 - 13 | 1723 | 104 | ||
BC151157.1 | - | 12 - 13 | 1684 | 9088045 | 104 | |
BC172024.1 | - | 12 - 13 | 1684 | 9054310 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |