Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007467.3 | NP_031493.2 | 1 - 2 | 260 | 70 | |
GenBank mRNA | AK154992.1 | - | 1 - 2 | 214 | 70 | |
AK160871.1 | - | 1 - 2 | 209 | 70 | ||
AK170818.1 | - | 1 - 2 | 193 | 70 | ||
BC021877.1 | - | 1 - 2 | 239 | 4481838 | 70 | |
BC106158.1 | - | 1 - 2 | 167 | 70 | ||
L04538.1 | - | 1 - 2 | 230 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus 1700 Probe spans exons |