Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007399.3 | NP_031425.2 | 2 - 3 | 412 | 86 | |
GenBank mRNA | AF011379.1 | - | 2 - 3 | 233 | 86 | |
BC057346.1 | - | 2 - 3 | 374 | 86 | ||
BC066207.1 | - | 2 - 3 | 417 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |