Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amotl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081395.1 | NP_001074864.1 | 9 - 10 | 2389 | 81 | |
GenBank mRNA | AK016526.1 | - | 10 - 11 | 2249 | 81 | |
AK220238.1 | - | 7 - 8 | 1623 | 81 | ||
AX763684.1 | - | 7 - 8 | 1913 | 81 | ||
BC055117.1 | - | 1 - 2 | 114 | 81 | ||
BC151000.1 | - | 9 - 10 | 2197 | 9087913 | 81 | |
BC151004.1 | - | 9 - 10 | 2197 | 9087917 | 81 | |
BC151007.1 | - | 9 - 10 | 2197 | 9087920 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |