Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eepd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026189.3 | NP_080465.3 | 4 - 5 | 1506 | 68 | |
GenBank mRNA | AK009180.2 | - | 4 - 5 | 1501 | 68 | |
AK168564.1 | - | 4 - 5 | 1507 | 68 | ||
AK173239.1 | - | 4 - 5 | 1556 | 68 | ||
BC025571.1 | - | 4 - 5 | 1570 | 5323841 | 68 | |
CK619108.1 | - | 3 - 4 | 234 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |