Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900022919.mm.1 |
NM_001160378.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022919.mm.1 |
NM_001160378.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022919.mm.1 |
TC0900002770.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022920.mm.1 |
NM_001160378.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022920.mm.1 |
NM_001160378.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022920.mm.1 |
NM_001160379.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022920.mm.1 |
NM_001160379.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022920.mm.1 |
TC0900002770.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022918.mm.1 |
NM_001160378.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022918.mm.1 |
NM_001160378.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022918.mm.1 |
NM_001160379.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022918.mm.1 |
NM_001160379.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022918.mm.1 |
TC0900002770.mm.1 |
MTA TC |