Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133718.4 | NP_598479.1 | 1 - 2 | 456 | 190 | |
GenBank mRNA | AK008408.1 | - | 1 - 2 | 342 | 190 | |
AK077065.1 | - | 1 - 2 | 459 | 190 | ||
AK150071.1 | - | 1 - 2 | 359 | 190 | ||
AK159364.1 | - | 1 - 2 | 383 | 190 | ||
AK161096.1 | - | 1 - 2 | 338 | 190 | ||
AK170171.1 | - | 1 - 2 | 359 | 190 | ||
AK170697.1 | - | 1 - 2 | 360 | 190 | ||
AK200429.1 | - | 1 - 2 | 120 | 190 | ||
BC018367.1 | - | 1 - 2 | 308 | 2648510 | 190 | |
BC018491.1 | - | 1 - 2 | 336 | 2651602 | 190 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |