Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cisd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134007.4 | NP_598768.1 | 2 - 3 | 339 | 69 | |
GenBank mRNA | AK075907.1 | - | 2 - 3 | 340 | 69 | |
AK131595.1 | - | 2 - 3 | 339 | 69 | ||
AK131605.1 | - | 2 - 3 | 318 | 69 | ||
AK131812.1 | - | 2 - 3 | 343 | 69 | ||
AK131995.1 | - | 2 - 3 | 350 | 69 | ||
AK167898.1 | - | 2 - 3 | 318 | 69 | ||
BC013522.1 | - | 2 - 3 | 328 | 4236502 | 69 | |
BC019860.1 | - | 2 - 3 | 355 | 5123692 | 69 | |
BC021952.1 | - | 2 - 3 | 376 | 5102315 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |