Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022653.4 | NP_073144.3 | 3 - 4 | 563 | 72 | |
GenBank mRNA | AF314187.1 | - | 3 - 4 | 521 | 72 | |
BC031722.1 | - | 3 - 4 | 529 | 4019169 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |