Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008688.3 | NP_032714.1 | 5 - 6 | 900 | 69 | |
NM_026756.2 | NP_081032.2 | 5 - 6 | 900 | 69 | ||
GenBank mRNA | AF358455.1 | - | 4 - 5 | 299 | 69 | |
AF358456.1 | - | 4 - 5 | 299 | 69 | ||
AF358457.1 | - | 4 - 5 | 299 | 69 | ||
AF358458.1 | - | 4 - 5 | 299 | 69 | ||
AK087539.1 | - | 5 - 6 | 885 | 69 | ||
AK147496.1 | - | 5 - 6 | 885 | 69 | ||
AK155258.1 | - | 5 - 6 | 902 | 69 | ||
U57635.1 | - | 5 - 6 | 840 | 69 | ||
Y07692.1 | - | 5 - 6 | 859 | 69 | ||
Y07693.1 | - | 5 - 6 | 1205 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |